3-Diethylamino-1-propyne ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N,N-Diethylpropargylamine
L12237-06
L12237-14
AAAL12237-06EA
50
USD
AAAL12237-06
AAAL12237-14
3-Diethylamino-1-propyne ≥98%
3-Diethylamino-1-propyne
Purity: 98+%
CAS No.: 4079-68-9
Molecular Formula: C7H13N
Formula Weight: 111.19
Flash Point: 20°(68°F)
Refractive Index: 1.4305
Odor: Amine-like
Boiling Point: 119-120°
Beilstein Reference: 1098468
UN No.: 2733
MDL No.: MFCD00039881
Formula:
C₇H₁₃N MW: 111.19 g/mol Boiling Pt: 119…120 °C Density: 0.804 Flash Pt: 20 °C(68 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00039881 CAS Number: 4079-68-9 EINECS: 223-804-5 UN: 2733 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...