Decyltrimethylammonium bromide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (1-Decyl)trimethylammonium bromide, N,N,N-Trimethyldecan-1-aminium bromide

L12144-09 L12144-18
AAAL12144-09EA 51.7 USD
AAAL12144-09 AAAL12144-18
Decyltrimethylammonium bromide 98%
Decyltrimethylammonium bromide

Purity: 98% CAS No.: 2082-84-0 Molecular Formula: CH3(CH2)9N(CH3)3Br Formula Weight: 280.30 Melting Point: 243-245° Beilstein Reference: 3915222

Formula: C₁₃H₃₀BrN
MW: 280.30 g/mol
Melting Pt: 243…245 °C
Storage Temperature: Ambient
MDL Number: MFCD00041973
CAS Number: 2082-84-0
EINECS: 218-219-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR