1-Adamantaneacetic acid ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(Adamantan-1-yl)acetic acid, 1-Carboxymethyladamantane
L12029-03
L12029-14
L12029-06
AAAL12029-03EA
42
USD
AAAL12029-03
AAAL12029-14
AAAL12029-06
1-Adamantaneacetic acid ≥98%
1-Adamantaneacetic acid
Purity: 98+%
CAS No.: 4942-47-6
Molecular Formula: C12H18O2
Formula Weight: 194.28
Melting Point: 136-138°
Beilstein Reference: 641412
MDL No.: MFCD00074728
Formula:
C₁₂H₁₈O₂ MW: 194.27 g/mol Melting Pt: 136…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00074728 CAS Number: 4942-47-6 EINECS: 225-585-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...