Dimethyl-trans-stilbene-4,4'-dicarboxylate ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: trans-Stilbene-4,4'-dicarboxylic Acid Dimethyl Ester

L11969-03 L11969-MD
AAAL11969-03EA 108 USD
AAAL11969-03 AAAL11969-MD
Dimethyl-trans-stilbene-4,4'-dicarboxylate ≥98%
Dimethyl-trans-stilbene-4,4'-dicarboxylate

Purity: 98+% CAS No.: 34541-73-6 Molecular Formula: C18H16O4 Formula Weight: 296.33 Melting Point: 234-236° Beilstein Reference: 2471189 MDL No.: MFCD00039527

Formula: C₁₈H₁₆O₄
MW: 296.32 g/mol
Melting Pt: 234…236 °C
Storage Temperature: Ambient
MDL Number: MFCD00039527
CAS Number: 34541-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR