Pentafluorophenyl isothiocyanate 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L11874-03
L11874-06
AAAL11874-03EA
89.4
USD
AAAL11874-03
AAAL11874-06
Pentafluorophenyl isothiocyanate 96%
Pentafluorophenyl isothiocyanate
Purity: 96%
CAS No.: 35923-79-6
Molecular Formula: C7F5NS
Formula Weight: 225.14
Storage Temperature: 2°C to 8°C
Refractive Index: 1.5378
Boiling Point: 109-110°/35mm
Beilstein Reference: 2736207
UN No.: 2922
MDL No.: MFCD00041042
Formula:
C₇F₅NS MW: 225.14 g/mol Boiling Pt: 109…110 °C/35 mm Density: 1.560 Storage Temperature: Refrigerator |
MDL Number:
MFCD00041042 CAS Number: 35923-79-6 EINECS: 252-791-9 UN: 2922 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...