Methyl-4,4,4-trifluorocrotonate 97%

Supplier: Thermo Scientific Chemicals
Warning

L11466-03 L11466-06
AAAL11466-03EA 59.2 USD
AAAL11466-03 AAAL11466-06
Methyl-4,4,4-trifluorocrotonate 97%
Methyl 4,4,4-trifluorocrotonate

Purity: 97% CAS No.: 85694-31-1 Molecular Formula: CF3CH=CHCO2CH3 Formula Weight: 154.10 Refractive Index: 1.3535 Boiling Point: 94-95° UN No.: 3272 MDL No.: MFCD00077567

Formula: C₅H₅F₃O₂
MW: 154.10
Boiling Pt: 94…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00077567
CAS Number: 85694-31-1
UN: 3272
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR