2-Ethynylpyridine 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with hydroquinone (HQ) 0.01%
Stab. with 0.01% hydroquinone
Purity: 98%
CAS No.: 1945-84-2
Molecular Formula: C7H5N
Formula Weight: 103.12
Storage Temperature: 2°C to 8°C
Flash Point: 76°(168°F)
Refractive Index: 1.5590
Odor: Characteristic
Boiling Point: 77-78°/14mm
Beilstein Reference: 1098904
MDL No.: MFCD00041598
Formula:
C₇H₅N Boiling Pt: 77…78 °C/14 mm Density: 1.020 Flash Pt: 76 °C(168 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041598 CAS Number: 1945-84-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...