(4-Nitrophenyl)-α-D-glucopyranoside 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Nitrophenyl α-D-glucopyranoside, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol, 4-Nitrophenyl a-D-glucopyranoside

L10070-06 L10070-03
AAAL10070-06EA 197 USD
AAAL10070-06 AAAL10070-03
(4-Nitrophenyl)-α-D-glucopyranoside 98+%
(4-Nitrophenyl)-α-D-glucopyranoside

Purity: 98+% CAS No.: 3767-28-0 Molecular Formula: C12H15NO8 Formula Weight: 301.25 Storage Temperature: 2°C to 8°C Melting Point: 206-207° Beilstein Reference: 92212 Optical Rotation: +218° (c=0.5 in water) MDL No.: MFCD00064088

Formula: C₁₂H₁₅NO₈
MW: 301.25 g/mol
Melting Pt: 206…207 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064088
CAS Number: 3767-28-0
EINECS: 223-189-3

Order Now

Specification Test Results

Optical rotation +218° (c=0.5 in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR