Methyl-2-acetamidoacrylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Acetamidoacrylic Acid Methyl Ester, Methyl 2-acetamidoacrylate
L10049-06
L10049-03
AAAL10049-06EA
175
USD
AAAL10049-06
AAAL10049-03
Methyl-2-acetamidoacrylate 97%
Methyl-2-acetamidoacrylate
Fieser: 17,46
Purity: 99%
CAS No.: 35356-70-8
Molecular Formula: C6H9NO3
Formula Weight: 143.14
Storage Temperature: 2°C to 8°C
Melting Point: 49-51°
Boiling Point: 103-104°/8mm
Beilstein Reference: 1812208
MDL No.: MFCD00013394
Formula:
C₆H₉NO₃ MW: 143.14 g/mol Boiling Pt: 103…104 °C (8 mmHg) Melting Pt: 49…51 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00013394 CAS Number: 35356-70-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...