2,1,3-Benzothiadiazol-4-ylamine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Aminobenzo-2,1,3-thiadiazole, 4-Aminopiazthiole, Benzo[c][1,2,5]thiadiazol-4-amine, 4-Amino-2,1,3-benzothiadiazole

L09678-03 L09678-06
AAAL09678-03EA 62.8 USD
AAAL09678-03 AAAL09678-06
2,1,3-Benzothiadiazol-4-ylamine 98%
2,1,3-Benzothiadiazol-4-ylamine

Purity: 98% CAS No.: 767-64-6 Molecular Formula: C6H5N3S Formula Weight: 151.19 Melting Point: 67-69° Beilstein Reference: 3551

Formula: C₆H₅N₃S
MW: 151.19 g/mol
Melting Pt: 67…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00005810
CAS Number: 767-64-6
EINECS: 212-186-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR