Ethyl-3-isocyanatobenzoate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-(Ethoxycarbonyl)phenyl isocyanate

L09495-03 L09495-06 L09495-14
AAAL09495-03EA 37.9 USD
AAAL09495-03 AAAL09495-06 AAAL09495-14
Ethyl-3-isocyanatobenzoate 97%
Ethyl 3-isocyanatobenzoate

Purity: 97% CAS No.: 67531-68-4 Molecular Formula: C10H9NO3 Formula Weight: 191.19 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5310 Boiling Point: 213° Beilstein Reference: 7636626 UN No.: 2206 MDL No.: MFCD00013865

Formula: C₁₀H₉NO₃
MW: 191.19 g/mol
Boiling Pt: 213 °C
Density: 1.154
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00013865
CAS Number: 67531-68-4
UN: 2206
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR