2-Naphthylacetonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

L09384-06 L09384-14
AAAL09384-06EA 69.5 USD
AAAL09384-06 AAAL09384-14
2-Naphthylacetonitrile 97%
2-Naphthylacetonitrile

Purity: 97% CAS No.: 7498-57-9 Molecular Formula: C12H9N Formula Weight: 167.21 Melting Point: 82-84° Boiling Point: 303° UN No.: 3439 MDL No.: MFCD00004122

Formula: C₁₂H₉N
MW: 167.21 g/mol
Boiling Pt: 303 °C
Melting Pt: 82…84 °C
Density: 1.092
Storage Temperature: Ambient
MDL Number: MFCD00004122
CAS Number: 7498-57-9
EINECS: 231-352-5
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR