5-Acetyl-2-amino-4-methylthiazole ≥97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(2-Amino-4-methylthiazol-5-yl)ethanone

L09108-14 L09108-06
AAAL09108-14EA 225 USD
AAAL09108-14 AAAL09108-06
5-Acetyl-2-amino-4-methylthiazole ≥97%
5-Acetyl-2-amino-4-methylthiazole

Purity: 97+% CAS No.: 30748-47-1 Molecular Formula: C6H8N2OS Formula Weight: 156.21 Melting Point: ca 268° dec. Beilstein Reference: 127111 MDL No.: MFCD00051952

Formula: C₆H₈N₂OS
MW: 156.21 g/mol
Melting Pt: ca 268 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00051952
CAS Number: 30748-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR