2-Ethoxy-4-methylphenol 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L08898-04
L08898-09
AAAL08898-04EA
73
USD
AAAL08898-04
AAAL08898-09
2-Ethoxy-4-methylphenol 95%
2-Ethoxy-4-methylphenol
Purity: 95%
CAS No.: 2563-07-7
Molecular Formula: C9H12O2
Formula Weight: 152.19
Refractive Index: 1.5230
Melting Point: 28-30°
Boiling Point: 103°/3mm
Beilstein Reference: 1865448
MDL No.: MFCD00043697
Formula:
C₉H₁₂O₂ MW: 152.19 g/mol Boiling Pt: 103 °C/3 mm Melting Pt: 28…30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043697 CAS Number: 2563-07-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...