D-(+)-Cellobiose octaacetate

Supplier: Thermo Scientific Chemicals

L08780-14 L08780-06
AAAL08780-14EA 131 USD
AAAL08780-14 AAAL08780-06
D-(+)-Cellobiose octaacetate
D(+)-Cellobiose octaacetate

CAS No.: 5346-90-7 Molecular Formula: C28H38O19 Formula Weight: 678.59 Melting Point: 224-231° Beilstein Reference: 79280 Merck Reference: 14,1961 Optical Rotation: +40° (c=1 in chloroform) MDL No.: MFCD00009600

Formula: C₂₈H₃₈O₁₉
MW: 678.59
Melting Pt: 224…231 °C
Storage Temperature: Ambient
MDL Number: MFCD00009600
CAS Number: 5346-90-7
EINECS: 226-304-5
Merck Index: 14,1961

Order Now

Specification Test Results

Optical rotation +40° (c=1 in chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR