Fluoral-P (=4-amino-3-penten-2-one) 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L08379-03
L08379-06
AAAL08379-03EA
28.82
USD
AAAL08379-03
AAAL08379-06
Fluoral-P (=4-amino-3-penten-2-one) 96%
Fluoral-P (=4-amino-3-penten-2-one)
Purity: 96%
CAS No.: 1118-66-7
Molecular Formula: C5H9NO
Formula Weight: 99.13
Melting Point: 36-41°
Boiling Point: 209-213°
Beilstein Reference: 1209344
MDL No.: MFCD00043715
Formula:
C₅H₉NO Boiling Pt: 209…213 °C Melting Pt: 36…41 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043715 CAS Number: 1118-66-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...