1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(3,5-Dimethyl-1H-pyrazol-1-yl)-3-oxopropanenitrile

L08369-06 L08369-14
AAAL08369-06EA 86.8 USD
AAAL08369-06 AAAL08369-14
1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole 97%
1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole

Purity: 97% CAS No.: 36140-83-7 Molecular Formula: C8H9N3O Formula Weight: 163.18 Melting Point: 118-122° Beilstein Reference: 6286 MDL No.: MFCD00051954

Formula: C₈H₉N₃O
MW: 163.18 g/mol
Melting Pt: 118…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00051954
CAS Number: 36140-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR