N6-Benzoyladenine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Benzamidopurine, N-(7H-Purin-6-yl)benzamide
L08292-06
L08292-14
L08292-03
AAAL08292-06EA
179
USD
AAAL08292-06
AAAL08292-14
AAAL08292-03
N6-Benzoyladenine 99%
N6-Benzoyladenine
Purity: 99%
CAS No.: 4005-49-6
Molecular Formula: C12H9N5O
Formula Weight: 239.23
Melting Point: 241-244°
Beilstein Reference: 20585
MDL No.: MFCD00037927
Formula:
C₁₂H₉N₅O MW: 239.24 g/mol Melting Pt: 241…244 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037927 CAS Number: 4005-49-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...