Methyl-β-D-galactopyranoside
Supplier: Thermo Scientific Chemicals
Synonyms:
Methyl-β-D-galactoside, (2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol, (2R,3R,4S,5R,6R)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol
L08127-03
L08127-06
AAAL08127-03EA
44.3
USD
AAAL08127-03
AAAL08127-06
Methyl-β-D-galactopyranoside
Methyl-beta-D-galactopyranoside
CAS No.: 1824-94-8
Molecular Formula: C7H14O6
Formula Weight: 194.18
Storage Temperature: 2°C to 8°C
Melting Point: 175-179°
Beilstein Reference: 81569
Optical Rotation: -16° (c=1.5 in methanol)
MDL No.: MFCD00064357
Formula:
C₇H₁₄O₆ Melting Pt: 175…179 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00064357 CAS Number: 1824-94-8 EINECS: 217-361-7 |
Specification Test Results
Optical rotation | -16° (c=1.5 in methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...