4-(Methylthio)-m-cresol 97%
Supplier: Thermo Scientific Chemicals
L07353-06
L07353-14
AAAL07353-06EA
36.2
USD
AAAL07353-06
AAAL07353-14
4-(Methylthio)-m-cresol 97%
4-(Methylthio)-m-cresol
Purity: 97%
CAS No.: 3120-74-9
Molecular Formula: C8H10OS
Formula Weight: 154.23
Melting Point: 54-58°
Beilstein Reference: 1210322
MDL No.: MFCD00045773
Formula:
C₈H₁₀OS |
MDL Number:
MFCD00045773 CAS Number: 3120-74-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...