3-Fluoro-DL-phenylalanine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
2-Amino-3-(3-fluorophenyl)propionic acid, H-DL-Phe(3-F)-OH
L07297-03
L07297-06
AAAL07297-03EA
43.1
USD
AAAL07297-03
AAAL07297-06
3-Fluoro-DL-phenylalanine 98%
3-Fluoro-DL-phenylalanine
Purity: 98%
CAS No.: 2629-54-1
Molecular Formula: C9H10FNO2
Formula Weight: 183.18
Melting Point: ca 245° dec.
Beilstein Reference: 2939807
Formula:
C₉H₁₀FNO₂ MW: 183.2 g/mol Melting Pt: ∼ 245 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00004273 CAS Number: 2629-54-1 EINECS: 220-104-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...