2-(2-Aminophenyl)benzimidazole 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-(1H-Benzimidazol-2-yl)aniline, 2-(2-Aminophenyl)-1H-benzimidazole

L07034-14 L07034-06
AAAL07034-14EA 158.4 USD
AAAL07034-14 AAAL07034-06
2-(2-Aminophenyl)benzimidazole 98%
2-(2-Aminophenyl)benzimidazole

Purity: 98% CAS No.: 5805-39-0 Molecular Formula: C13H11N3 Formula Weight: 209.24 Melting Point: 212-216° Beilstein Reference: 173018 MDL No.: MFCD00051496

Formula: C₁₃H₁₁N₃
MW: 209.252 g/mol
Melting Pt: 212…216 °C
Storage Temperature: Ambient
MDL Number: MFCD00051496
CAS Number: 5805-39-0
EINECS: 227-361-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR