3,3-Dimethyl-1,2-epoxybutane 95%

Supplier: Thermo Scientific Chemicals
Danger

L06951-14 L06951-06
AAAL06951-14EA 600 USD
AAAL06951-14 AAL06951-06
3,3-Dimethyl-1,2-epoxybutane 95%
3,3-Dimethyl-1,2-epoxybutane

Purity: 95% CAS No.: 2245-30-9 Molecular Formula: C6H12O Formula Weight: 100.16 Flash Point: -1°(30°F) Refractive Index: 1.4010 Odor: Recognizable Boiling Point: 95-96° Beilstein Reference: 102478 UN No.: 1993 MDL No.: MFCD00051590

Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 95…96 °C
Density: 0.822
Flash Pt: -1 °C(30 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051590
CAS Number: 2245-30-9
EINECS: 218-831-4
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR