3-Phenylpropionitrile 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hydrocinnamonitrile

L06483-09 L06483-18
AAAL06483-09EA 52.3 USD
AAAL06483-09 AAAL06483-18
3-Phenylpropionitrile 98%
3-Phenylpropionitrile

Purity: 98% CAS No.: 645-59-0 Molecular Formula: C6H5(CH2)2CN Formula Weight: 131.18 Flash Point: 110°(230°F) Refractive Index: 1.5210 Melting Point: -2° Boiling Point: 261° Beilstein Reference: 636348 UN No.: 3276 MDL No.: MFCD00001961

Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 261 °C
Melting Pt: –2 °C
Density: 1.008
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001961
CAS Number: 645-59-0
EINECS: 211-447-8
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR