4-Ethoxycinnamic acid predominantly trans ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(4-Ethoxyphenyl)acrylic Acid
L06324-06
L06324-14
AAAL06324-06EA
80
USD
AAAL06324-06
AAAL06324-14
4-Ethoxycinnamic acid predominantly trans ≥98%
4-Ethoxycinnamic acid
Purity: 98+%
CAS No.: 2373-79-7
Molecular Formula: C11H12O3
Formula Weight: 192.22
Melting Point: 195-199°
Beilstein Reference: 3198068
MDL No.: MFCD00016848
Formula:
C₁₁H₁₂O₃ MW: 192.21 g/mol Melting Pt: 195…199 °C Storage Temperature: Ambient |
MDL Number:
MFCD11040581 CAS Number: 2373-79-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...