5-Chlorothiazol-2-amine hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (5-Chlorothiazol-2-yl)amine hydrochloride, 2-Amino-5-chlorothiazole hydrochloride

L05707-06 L05707-14 L05707-03
AAAL05707-06EA 95 USD
AAAL05707-06 AAAL05707-14 AAAL05707-03
5-Chlorothiazol-2-amine hydrochloride 97%
5-Chlorothiazol-2-amine hydrochloride

Purity: 97% CAS No.: 55506-37-1 Molecular Formula: C3H3ClN2S.HCl Formula Weight: 171.05 Melting Point: 168-172° MDL No.: MFCD00039006

Formula: C3H4Cl2N2S
MW: 171.05 g/mol
Melting Pt: 168…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00039006
CAS Number: 55506-37-1
EINECS: 259-680-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR