N,N-Dimethylbutyramide 98%
Supplier: Thermo Scientific Chemicals
L05556-06
L05556-14
AAAL05556-06EA
69.5
USD
AAAL05556-06
AAAL05556-14
N,N-Dimethylbutyramide 98%
N,N-Dimethylbutyramide
Purity: 98%
CAS No.: 760-79-2
Molecular Formula: C6H13NO
Formula Weight: 115.17
Refractive Index: 1.4425
Boiling Point: 80-82°/15mm
Beilstein Reference: 1742172
MDL No.: MFCD00015222
Formula:
C₆H₁₃NO MW: 115.17 g/mol Boiling Pt: 80…82 °C/15 mm Storage Temperature: Ambient |
MDL Number:
MFCD00015222 CAS Number: 760-79-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...