9-Aminofluorene hydrochloride ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: 9-Fluorenamine Hydrochloride, 9H-Fluoren-9-amine hydrochloride

L05156-03 L05156-06
AAAL05156-03EA 26.5 USD
AAAL05156-03 AAAL05156-06
9-Aminofluorene hydrochloride ≥98%
9-Aminofluorene hydrochloride

Purity: 98+% CAS No.: 5978-75-6 Molecular Formula: C13H11N.HCl Formula Weight: 217.70 Melting Point: ca 225° Beilstein Reference: 3915368 MDL No.: MFCD00012536

Formula: C₁₃H₁₁N
MW: 217.70 g/mol
Melting Pt: ∼ 225 °C
Storage Temperature: Ambient
MDL Number: MFCD00012536
CAS Number: 5978-75-6
EINECS: 227-778-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR