3-Bromo-2,4,6-trimethylnitrobenzene ≥90%

Supplier: Thermo Scientific Chemicals
Warning

L05107-06 L05107-03
AAAL05107-06EA 216 USD
AAAL05107-06 AAAL05107-03
3-Bromo-2,4,6-trimethylnitrobenzene ≥90%
3-Bromo-2,4,6-trimethylnitrobenzene

Purity: 90+% CAS No.: 90561-85-6 Molecular Formula: C9H10BrNO2 Formula Weight: 244.09 Melting Point: 56-62° Boiling Point: 106-107°/1.3mm Beilstein Reference: 1958703 MDL No.: MFCD00015924

Formula: C₉H₁₀BrNO₂
Boiling Pt: 106…107 °C/1.3 mm
Melting Pt: 56…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00015924
CAS Number: 90561-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR