4-Isopropylphenylacetic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(4-Isopropylphenyl)acetic acid

L05071-03 L05071-06
AAAL05071-03EA 80.1 USD
AAAL05071-03 AAAL05071-06
4-Isopropylphenylacetic acid 98+%
4-Isopropylphenylacetic acid

Purity: 98+% CAS No.: 4476-28-2 Molecular Formula: C11H14O2 Formula Weight: 178.23 Melting Point: 51-52° Beilstein Reference: 2328756

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Melting Pt: 51…52 °C
Storage Temperature: Ambient
MDL Number: MFCD00041035
CAS Number: 4476-28-2
EINECS: 224-755-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR