n-Butyl phenyl ether 99%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
N-butylphenylether
L04703-22
L04703-14
AAAL04703-22EA
108
USD
AAAL04703-22
AAAL04703-14
n-Butyl phenyl ether 99%
n-Butyl phenyl ether
Purity: 99%
CAS No.: 1126-79-0
Molecular Formula: C6H5OCH2CH2CH2CH3
Formula Weight: 150.22
Flash Point: 82°(179°F)
Refractive Index: 1.4970
Melting Point: -19°
Boiling Point: 210-211°
Beilstein Reference: 1635559
MDL No.: MFCD00009438
Formula:
C₁₀H₁₄O Boiling Pt: 210…211 °C Melting Pt: –19 °C Density: 0,938 Flash Pt: 82 °C(179 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009438 CAS Number: 1126-79-0 EINECS: 214-426-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...