Ethyl hydrogen suberate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
8-Ethoxy-8-oxooctanoic acid
L04272-06
L04272-14
AAAL04272-06EA
86.3
USD
AAAL04272-06
AAAL04272-14
Ethyl hydrogen suberate 98%
Ethyl hydrogen suberate
Purity: 98%
CAS No.: 14113-01-0
Molecular Formula: CH3CH2O2C(CH2)6CO2H
Formula Weight: 202.25
Melting Point: 25-27°
Beilstein Reference: 1775737
MDL No.: MFCD00014385
Formula:
CH₃CH₂O₂C(CH₂)₆CO₂H MW: 202.25 g/mol Melting Pt: 25…27 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014385 CAS Number: 14113-01-0 EINECS: 237-968-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...