Vinyl(chloromethyl)dimethylsilane 97%

Supplier: Thermo Scientific Chemicals
Danger

L04056-06 L04056-14
AAAL04056-06EA 64.2 USD
AAAL04056-06 AAAL04056-14
Vinyl(chloromethyl)dimethylsilane 97%
Vinyl(chloromethyl)dimethylsilane

Purity: 97% CAS No.: 16709-86-7 Molecular Formula: C5H11ClSi Formula Weight: 134.68 Flash Point: 14°(57°F) Refractive Index: 1.4430 Boiling Point: 121-122° Beilstein Reference: 2071374 UN No.: 1993 MDL No.: MFCD00040860

Formula: C₅H₁₁ClSi
MW: 134.68 g/mol
Boiling Pt: 121…122 °C
Density: 0.893
Flash Pt: 14 °C(57 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040860
CAS Number: 16709-86-7
EINECS: 240-758-1
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR