Diallyl maleate 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Diallyl Maleate
L03867-30
L03867-0B
AAAL03867-18EA
46.86
USD
AAAL03867-18
AAAL03867-30
AAAL03867-0B
Diallyl maleate 95%
Diallyl maleate
Purity: 95%
CAS No.: 999-21-3
Molecular Formula: C10H12O4
Formula Weight: 196.21
Flash Point: 130°(266°F)
Refractive Index: 1.4670
Melting Point: -47°
Boiling Point: 109-110°/3mm
Beilstein Reference: 1725954
UN No.: 2810
MDL No.: MFCD00010763
Formula:
C₁₀H₁₂O₄ Boiling Pt: 109…110 °C/3 mm Melting Pt: –47 °C Density: 1.072 Flash Pt: 130 °C(266 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010763 CAS Number: 999-21-3 EINECS: 213-658-0 UN: 2810 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...