Methyl Phenylsulfonylacetate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Phenylsulphonylacetic Acid Methyl Ester

L03748-06 L03748-14
AAAL03748-06EA 29.6 USD
AAAL03748-06 AAAL03748-14
Methyl Phenylsulfonylacetate 98%
Methyl Phenylsulfonylacetate

Purity: 98% CAS No.: 34097-60-4 Molecular Formula: C9H10O4S Formula Weight: 214.24 Flash Point: 110°(230°F) Refractive Index: 1.5350 Melting Point: ca 30° Boiling Point: 165°/0.05mm Beilstein Reference: 2051463 MDL No.: MFCD00007555

Formula: C₉H₁₀O₄S
MW: 214.24 g/mol
Boiling Pt: 165 °C/0.05 mm
Melting Pt: ca 30 °C
Density: 1.240
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007555
CAS Number: 34097-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR