Cyclohexyl methyl sulfide 97%

Supplier: Thermo Scientific Chemicals
Warning

L03712-14 L03712-06
AAAL03712-14EA 116 USD
AAAL03712-14 AAAL03712-06
Cyclohexyl methyl sulfide 97%
Cyclohexyl methyl sulphide

Purity: 97% CAS No.: 7133-37-1 Molecular Formula: C7H14S Formula Weight: 130.25 Flash Point: 61°(142°F) Refractive Index: 1.4940 Odor: Unpleasant Melting Point: -80° Boiling Point: 89-90°/35mm Beilstein Reference: 1919309 UN No.: 3334 MDL No.: MFCD00039460

Formula: C₇H₁₄S
MW: 130.25 g/mol
Boiling Pt: 89…90 °C/35 mm
Melting Pt: –80 °C
Density: 0.944
Flash Pt: 61 °C(142 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039460
CAS Number: 7133-37-1
EINECS: 230-431-1
UN: 3334
ADR: "9",

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR