3,5-Dimethyl-4-nitrobenzoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L02704-03
L02704-06
AAAL02704-03EA
208
USD
AAAL02704-03
AAAL02704-06
3,5-Dimethyl-4-nitrobenzoic acid 97%
3,5-Dimethyl-4-nitrobenzoic acid
Purity: 97%
CAS No.: 3095-38-3
Molecular Formula: C9H9NO4
Formula Weight: 195.18
Odor: Odorless
Melting Point: 222-226°
Beilstein Reference: 1965772
MDL No.: MFCD00014699
Formula:
C₉H₉NO₄ Melting Pt: 222…226 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014699 CAS Number: 3095-38-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...