2,2'-Dinaphthyl ether ≥98%
Supplier: Thermo Scientific Chemicals
L02565-06
L02565-14
AAAL02565-06EA
64.2
USD
AAAL02565-06
AAAL02565-14
2,2'-Dinaphthyl ether ≥98%
2,2'-Dinaphthyl ether
Purity: 98+%
CAS No.: 613-80-9
Molecular Formula: C20H14O
Formula Weight: 270.33
Melting Point: 103-105°
Beilstein Reference: 2054097
MDL No.: MFCD00015447
Formula:
C₂₀H₁₄O Melting Pt: 103…105 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015447 CAS Number: 613-80-9 EINECS: 210-356-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...