Benzofuran-2-carboxaldehyde 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Benzo[b]furan-2-carboxaldehyde, Benzofuran-2-carbaldehyde
L02456-04
L02456-09
AAAL02456-04EA
113.63
USD
AAAL02456-04
AAAL02456-09
Benzofuran-2-carboxaldehyde 99%
Benzofuran-2-carboxaldehyde
Purity: 99%
CAS No.: 4265-16-1
Molecular Formula: C9H6O2
Formula Weight: 146.15
Storage Temperature: 2°C to 8°C
Refractive Index: 1.6320
Boiling Point: 92-97°/1mm
Beilstein Reference: 2910
MDL No.: MFCD00015463
Formula:
C₉H₆O₂ MW: 146.145 g/mol Boiling Pt: 92…97 °C/1 mm Storage Temperature: Ambient |
MDL Number:
MFCD00015463 CAS Number: 4265-16-1 EINECS: 224-248-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...