Dicyclopropylmethanol 97%

Supplier: Thermo Scientific Chemicals

L01816-03 L01816-06
AAAL01816-03EA 38.8 USD
AAAL01816-03 AAAL01816-06
Dicyclopropylmethanol 97%
Dicyclopropylmethanol

Purity: 97% CAS No.: 14300-33-5 Molecular Formula: C7H12O Formula Weight: 112.17 Flash Point: 110°(230°F) Refractive Index: 1.4650 Boiling Point: 99-100°/12mm Beilstein Reference: 2203215 MDL No.: MFCD00019249

Formula: C₇H₁₂O
MW: 112.17 g/mol
Boiling Pt: 99…100 °C/12 mm
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00019249
CAS Number: 14300-33-5
EINECS: 238-236-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR