1-Naphthalenemethanol 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
Naphthalen-1-ylmethanol
L01347-09
L01347-18
AAAL01347-09EA
54.6
USD
AAAL01347-09
AAAL01347-18
1-Naphthalenemethanol 98+%
1-Naphthalenemethanol
Purity: 98+%
CAS No.: 4780-79-4
Molecular Formula: C11H10O
Formula Weight: 158.20
Melting Point: 58-63°
Boiling Point: 301°/715mm
MDL No.: MFCD00004044
Formula:
C₁₁H₁₀O MW: 158.20 g/mol Boiling Pt: 301 °C/715 mm Melting Pt: 58…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004044 CAS Number: 4780-79-4 EINECS: 225-324-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...