2,2'-Bipyrimidyl 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L01190-03
L01190-MD
AAAL01190-03EA
289
USD
AAAL01190-03
AAAL01190-MD
2,2'-Bipyrimidyl 96%
2,2'-Bipyrimidyl
Purity: 96%
CAS No.: 34671-83-5
Molecular Formula: C8H6N4
Formula Weight: 158.16
Melting Point: 110-116°
Beilstein Reference: 607396
MDL No.: MFCD00014600
Formula:
C₈H₆N₄ Melting Pt: 110…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014600 CAS Number: 34671-83-5 EINECS: 252-137-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...