N-tert-Butoxycarbonylimidazole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
tert-Butyl 1H-imidazole-1-carboxylate, N-Boc-imidazole
L00506-06
L00506-14
AAAL00506-06EA
29.1
USD
AAAL00506-06
AAAL00506-14
N-tert-Butoxycarbonylimidazole 98%
N-tert-Butoxycarbonylimidazole
Fieser: 1,98
Purity: 98%
CAS No.: 49761-82-2
Molecular Formula: C8H12N2O2
Formula Weight: 168.20
Storage Temperature: 2°C to 8°C
Melting Point: 44-48°
Beilstein Reference: 607792
MDL No.: MFCD00014497
Formula:
C₈H₁₂N₂O₂ MW: 168.20 g/mol Melting Pt: 44…48 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00014497 CAS Number: 49761-82-2 EINECS: 256-475-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...