Tetraphenylphosphine imide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L00496-03
L00496-06
AAAL00496-03EA
43.8
USD
AAAL00496-03
AAAL00496-06
Tetraphenylphosphine imide 97%
Tetraphenylphosphine imide
Purity: 97%
CAS No.: 2325-27-1
Molecular Formula: C24H20NP
Formula Weight: 353.40
Odor: Odorless
Melting Point: 128-132°
Beilstein Reference: 753032
MDL No.: MFCD00003044
Formula:
C₂₄H₂₀NP Melting Pt: 128…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003044 CAS Number: 2325-27-1 EINECS: 219-039-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...