Phenyl formate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L00289-14
L00289-06
AAAL00289-14EA
227
USD
AAAL00289-14
AAAL00289-06
Phenyl formate 95%
Phenyl formate
Purity: 95%
CAS No.: 1864-94-4
Molecular Formula: HCOOC6H5
Formula Weight: 122.12
Flash Point: 71°(159°F)
Refractive Index: 1.5120
Boiling Point: 72-74°/13mm
Beilstein Reference: 507113
MDL No.: MFCD00014126
Formula:
C₇H₆O₂ Boiling Pt: 72…74 °C (13 mmHg) Density: 1.090 Flash Pt: 71 °C (159 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014126 CAS Number: 1864-94-4 EINECS: 217-471-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...