2-Methyl-2-(4-methylphenyl)morpholine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Methyl-2-(p-tolyl)morpholine
H50732-03
H50732-MD
AAAH50732-03EA
280
USD
AAAH50732-03
AAAH50732-MD
2-Methyl-2-(4-methylphenyl)morpholine 97%
2-Methyl-2-(4-methylphenyl)morpholine
Purity: 97%
CAS No.: 902836-81-1
Molecular Formula: C12H17NO
Formula Weight: 191.28
Melting Point: 49-51°
MDL No.: MFCD08060961
Formula:
C₁₂H₁₇NO MW: 191.28 g/mol Melting Pt: 49…51 °C Storage Temperature: Ambient |
MDL Number:
MFCD08060961 CAS Number: 902836-81-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...