(3,5-Dichlorophenyl)methanesulfonyl chloride 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3,5-Dichloro-α-toluenesulphonyl chloride
H50707-03
H50707-MD
AAAH50707-03EA
145
USD
AAAH50707-03
AAAH50707-MD
(3,5-Dichlorophenyl)methanesulfonyl chloride 96%
(3,5-Dichlorophenyl)methanesulphonyl chloride
Purity: 96%
CAS No.: 163295-70-3
Molecular Formula: C7H5Cl3O2S
Formula Weight: 259.54
UN No.: 3096
MDL No.: MFCD01631928
Formula:
C₇H₅CI₃O₂S MW: 259.54 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01631928 CAS Number: 163295-70-3 UN: 3261 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...