4-Chloro-8-fluoroquinoline

Supplier: Thermo Scientific Chemicals
Warning

H50541-03 H50541-06
AAAH50541-03EA 126 USD
AAAH50541-03 AAAH50541-06
4-Chloro-8-fluoroquinoline
4-Chloro-8-fluoroquinoline

CAS No.: 63010-72-0 Molecular Formula: C9H5ClFN Formula Weight: 181.60 MDL No.: MFCD00278785

Formula: C₉H₅ClFN
MW: 181.60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00278785
CAS Number: 63010-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR