4-(1-Pyrrolidinyl)-1-butylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(Pyrrolidin-1-yl)butan-1-amine

H50323-06 H50323-03
AAAH50323-06EA 290 USD
AAAH50323-06 AAAH50323-03
4-(1-Pyrrolidinyl)-1-butylamine 98%
4-(1-Pyrrolidinyl)-1-butylamine

Purity: 98% CAS No.: 24715-90-0 Molecular Formula: C8H18N2 Formula Weight: 142.25 UN No.: 2735 MDL No.: MFCD00192591

Formula: C₈H₁₈N₂
MW: 142.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00192591
CAS Number: 24715-90-0
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR