4-(Diethylamino)butylamine 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H50278-03
H50278-06
AAAH50278-03EA
104
USD
AAAH50278-03
AAAH50278-06
4-(Diethylamino)butylamine 96%
4-(Diethylamino)butylamine
Purity: 96%
CAS No.: 27431-62-5
Molecular Formula: C8H20N2
Formula Weight: 144.26
Boiling Point: 72-24°/10mm
UN No.: 2735
MDL No.: MFCD00014836
Formula:
C₈H₂₀N₂ Boiling Pt: 72…24 °C/10 mm Density: 0.83 Storage Temperature: Ambient |
MDL Number:
MFCD00014836 CAS Number: 27431-62-5 EINECS: 248-456-1 UN: 2735 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...